Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VE-Cadherin Antibody (MM0012-8A03), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB11060978APC
Description
VE-Cadherin Monoclonal antibody specifically detects VE-Cadherin in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| VE-Cadherin | |
| Monoclonal | |
| APC | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| MM0012-8A03 | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| 7B4, 7B4 antigen, cadherin 5, type 2 (vascular endothelium), cadherin 5, type 2, VE-cadherin (vascular epithelium), cadherin-5, CD144, CD144 antigen, endothelial-specific cadherin, FLJ17376, Vascular endothelial cadherin, VE-cadherin | |
| Human recombinant VE-Cadherin (extracellular domain) | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication, Cellular Markers, Extracellular Matrix, Phospho Specific, Plasma Membrane Markers, Signal Transduction | |
| 1003 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction