Learn More
Invitrogen™ VE-cadherin Monoclonal Antibody (3D5C7)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA517050
Description
MA5-17050 targets CDH5 in FACS, indirect ELISA, and WB applications and shows reactivity with Human and Rat samples. The MA5-17050 immunogen is purified recombinant fragment of human CDH5 (amino acids: 29-223 ) expressed in E. Coli. MA5-17050 detects CDH5 which has a predicted molecular weight of approximately 87.5kDa.
VE-cadherin is a member of the cadherin superfamily that is located in a six-cadherin cluster in a region on the long arm of chromosome 16 and is involved in loss of heterozygosity events in breast and prostate cancer. VE-cadherin protein is a calcium-dependent cell-cell adhesion glycoprotein comprised of five extracellular cadherin repeats, a transmembrane region and a highly conserved cytoplasmic tail. Functioning as a classic cadherin by imparting to cells the ability to adhere in a homophilic manner, VE-cadherin may play an important role in endothelial cell biology through control of the cohesion and organization of the intercellular junctions. An alternative splice variant has been described but the full length sequence of VE-cadherin has not been determined.
Specifications
VE-cadherin | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
P33151 | |
CDH5 | |
Purified recombinant fragment of human CDH5 (amino acids: 29-223 ) expressed in E. Coli. | |
100 μg | |
Primary | |
Human, Rat | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Western Blot | |
3D5C7 | |
Unconjugated | |
CDH5 | |
7B4; 7B4 antigen; 7B4/cadherin-5; AA408225; cadherin 5; cadherin 5, type 2 (vascular endothelium); cadherin 5, type 2, VE-cadherin (vascular endothelium); cadherin 5, type 2, VE-cadherin (vascular epithelium); cadherin-5; cadherin-5; cd144 antigen; VE cadherin; Cd144; cd144 antigen; CDH5; cell-cell adhesion protein; endothelial-specific cadherin; vascular endothelial cadherin; vascular endothelial cadherin precursor; Vec; VEcad; VE-Cad; VE-cadherin; VECD | |
Mouse | |
Protein G | |
RUO | |
1003, 307618 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.