Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VE-cadherin Monoclonal Antibody (BV9), FITC, Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA549025
Description
VE-cadherin is a member of the cadherin superfamily that is located in a six-cadherin cluster in a region on the long arm of chromosome 16 and is involved in loss of heterozygosity events in breast and prostate cancer. VE-cadherin protein is a calcium-dependent cell-cell adhesion glycoprotein comprised of five extracellular cadherin repeats, a transmembrane region and a highly conserved cytoplasmic tail. Functioning as a classic cadherin by imparting to cells the ability to adhere in a homophilic manner, VE-cadherin may play an important role in endothelial cell biology through control of the cohesion and organization of the intercellular junctions. An alternative splice variant has been described but the full length sequence of VE-cadherin has not been determined.
Specifications
| VE-cadherin | |
| Monoclonal | |
| 100 μg/mL | |
| PBS with 1% BSA and 0.02% sodium azide | |
| P33151 | |
| CDH5 | |
| The monoclonal antibody BV9 binds to the extracellular domain (EC3-EC4) of human VE-cadherin (vascular endothelial cadherin) | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, Functional Assay, Immunocytochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, Inhibition Assays, Western Blot | |
| BV9 | |
| FITC | |
| CDH5 | |
| 7B4; 7B4 antigen; 7B4/cadherin-5; AA408225; cadherin 5; cadherin 5, type 2 (vascular endothelium); cadherin 5, type 2, VE-cadherin (vascular endothelium); cadherin 5, type 2, VE-cadherin (vascular epithelium); cadherin-5; cadherin-5; cd144 antigen; VE cadherin; Cd144; cd144 antigen; CDH5; cell-cell adhesion protein; endothelial-specific cadherin; vascular endothelial cadherin; vascular endothelial cadherin precursor; Vec; VEcad; VE-Cad; VE-cadherin; VECD | |
| Mouse | |
| Protein G | |
| RUO | |
| 1003 | |
| 4° C, store in dark | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction