Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VE-Cadherin, p Tyr731 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$208.00 - $755.00
Specifications
Concentration | 1 mg/mL |
---|---|
Dilution | Western Blot 1:500-1:1000 |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP254766
![]() |
Novus Biologicals
NBP254766 |
0.1 mg |
Each for $755.00
|
|
|||||
NBP25476605
![]() |
Novus Biologicals
NBP2547660.05MG |
0.05 mg |
Each for $474.50
|
|
|||||
NB413815
![]() |
Novus Biologicals
NBP2547660.025MG |
0.025 mg |
Each for $208.00
|
|
|||||
Description
VE-Cadherin Polyclonal specifically detects VE-Cadherin in Human, Mouse samples. It is validated for Western Blot.Specifications
1 mg/mL | |
Western Blot | |
Unconjugated | |
Human, Mouse | |
1003 | |
VE-Cadherin (phospho Tyr731) antibody was raised against a peptide sequence around phosphorylation site of Tyrosine731 (H-I-Y (p)-G-Y) derived from Human VE-Cadherin. | |
Primary |
Western Blot 1:500-1:1000 | |
Polyclonal | |
Rabbit | |
7B4, 7B4 antigen, cadherin 5, type 2 (vascular endothelium), cadherin 5, type 2, VE-cadherin (vascular epithelium), cadherin-5, CD144, CD144 antigen, endothelial-specific cadherin, FLJ17376, Vascular endothelial cadherin, VE-cadherin | |
CDH5 | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title