Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VE-Cadherin, p Tyr731 Antibody, mFluor Violet 500 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP254766MFV500
Description
VE-Cadherin Polyclonal antibody specifically detects VE-Cadherin in Human, Mouse samples. It is validated for Western BlotSpecifications
| VE-Cadherin | |
| Polyclonal | |
| Western Blot | |
| 7B4, 7B4 antigen, cadherin 5, type 2 (vascular endothelium), cadherin 5, type 2, VE-cadherin (vascular epithelium), cadherin-5, CD144, CD144 antigen, endothelial-specific cadherin, FLJ17376, Vascular endothelial cadherin, VE-cadherin | |
| VE-Cadherin (phospho Tyr731) antibody was raised against a peptide sequence around phosphorylation site of Tyrosine731 (H-I-Y (p)-G-Y) derived from Human VE-Cadherin. | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication, Cellular Markers, Extracellular Matrix, Phospho Specific, Plasma Membrane Markers, Signal Transduction | |
| 1003 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Epitope affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction