Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VE-Statin/EGFL7 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$170.97 - $492.89
Specifications
| Antigen | VE-Statin/EGFL7 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 1.0 mg/ml., Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB018646
![]() |
Novus Biologicals
NBP1471930.02MG |
0.02 mg |
Each for $170.97
|
|
|||||
NBP14719301
![]() |
Novus Biologicals
NBP1471930.1MG |
0.1 mg |
Each for $492.89
|
|
|||||
Description
VE-Statin/EGFL7 Polyclonal specifically detects VE-Statin/EGFL7 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| VE-Statin/EGFL7 | |
| Western Blot 1.0 mg/ml., Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| EGF-like protein 7, EGF-like-domain, multiple 7, epidermal growth factor-like protein 7, MEGF7, MGC111117, Multiple EGF-like domains protein 7, Multiple epidermal growth factor-like domains protein 7, NEU1 protein, NOTCH4-like protein, RP11-251M1.2, Vascular endothelial statin, VE-STATIN, Zneu1 | |
| EGFL7 | |
| IgG | |
| Protein A purified | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| LYOPH | |
| Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Angiogenesis | |
| PBS (pH 7.2) with No Preservative | |
| 51162 | |
| Recombinant human EGFL7 | |
| Primary | |
| Reconstitute in sterile water to a concentration of 0.1-1.0mg/mL. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title