Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VE-Statin/EGFL7 Antibody (OTI2B5), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274846AF350
Description
VE-Statin/EGFL7 Monoclonal specifically detects VE-Statin/EGFL7 in Human samples. It is validated for Western Blot.Specifications
VE-Statin/EGFL7 | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
EGFL7 | |
Recombinant protein fragment corresponding to amino acids 24-273 of human VE-Statin/EGFL7 (NP_057299) produced in E.coli. | |
Primary | |
Human | |
IgG1 |
Western Blot | |
OTI2B5 | |
Western Blot | |
EGF-like protein 7, EGF-like-domain, multiple 7, epidermal growth factor-like protein 7, MEGF7, MGC111117, Multiple EGF-like domains protein 7, Multiple epidermal growth factor-like domains protein 7, NEU1 protein, NOTCH4-like protein, RP11-251M1.2, Vascular endothelial statin, VE-STATIN, Zneu1 | |
Mouse | |
0.1 mL | |
51162 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction