Learn More
R&D Systems™ VectaCell Trolox Antifade Reagent for Live Cell Imaging
Shop All Novus Biologicals Products

Specifications
Specifications
| Product Line | Novus |
| For Use With (Application) | Immunocytochemistry/ Immunofluorescence, Cellular Imaging |
| Content And Storage | Store at 4°C. |
| Quantity | 2 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.