Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VEGF 121 Antibody (064), Alexa Fluor™ 488, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289193AF488
Description
VEGF 121 Monoclonal specifically detects VEGF 121 in Human samples. It is validated for ELISA, Sandwich ELISA Detection.Specifications
VEGF 121 | |
Monoclonal | |
Alexa Fluor 488 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Has cross-reactivity in ELISA with: Human VEGF165 No cross-reactivity in ELISA with: Human VEGFC Human VEGFD | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
64 | |
ELISA, Sandwich ELISA Detection | |
MVCD1, vascular endothelial growth factor A, Vascular permeability factor, VEGF-A, VEGFMGC70609, VPFvascular endothelial growth factor | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human VEGF 121 (P15692-9; Met1-Arg147). | |
0.1 mL | |
Angiogenesis, Biologically Active Proteins | |
7422 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction