Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VEGF 121 Antibody Pair (HRP), Novus Biologicals™
SDP

Catalog No. NB003003 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
15 Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB003003 15 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB003003 Supplier R&D Systems Supplier No. NBP27884315PLATES
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Monoclonal Antibody

VEGF 121 Monoclonal specifically detects VEGF 121 in Human samples. It is validated for ELISA/Sandwich ELISA.

Specifications

Antigen VEGF 121
Applications ELISA, Sandwich ELISA
Classification Monoclonal
Conjugate HRP
Dilution Sandwich ELISA
Gene Alias MVCD1, vascular endothelial growth factor A, Vascular permeability factor, VEGF-A, VEGFMGC70609, VPFvascular endothelial growth factor
Gene Symbols VEGFA
Host Species Rabbit
Quantity 15 Plates
Research Discipline Angiogenesis, Biologically Active Proteins
Gene ID (Entrez) 7422
Test Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human VEGF 121.
Target Species Human
Content And Storage Storage of components varies. See protocol for specific instructions.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.