Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ VEGF-164 Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA547021
Description
In sandwich ELISAs, less than 0.04% cross-reactivity with recombinant human (rh) VEGF121, rhVEGF165, and rhVEGF/PlGF is observed. Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
VEGF-164 is a gene target that encodes for the vascular endothelial growth factor. This protein is responsible for promoting angiogenesis. VEGF-164 has been shown to be involved in the development of various diseases, including cancer, cardiovascular disease, and diabetic retinopathy. Targeting this gene has become an important strategy in the development of new therapeutics for these conditions.
Specifications
VEGF-164 | |
Polyclonal | |
Unconjugated | |
VEGFA | |
Vascular Endothelial Growth Factor, Vascular Permeability Factor, VEGFA, VPF | |
Goat | |
Antigen affinity chromatography | |
RUO | |
22339 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
ELISA, Immunohistochemistry (Frozen), Neutralization, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q00731 | |
VEGFA | |
Sf 21-derived recombinant mouse VEGF164 Ala27-Arg190. | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction