Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VEGF 165 Antibody (105), mFluor Violet 450 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289780MFV450
Description
VEGF 165 Monoclonal antibody specifically detects VEGF 165 in Human, Cynomolgus Monkey samples. It is validated for ELISASpecifications
VEGF 165 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human, Cynomolgus Monkey | |
Purified |
ELISA | |
105 | |
ELISA | |
MVCD1, vascular endothelial growth factor A, Vascular permeability factor, VEGF-A, VEGFMGC70609, VPFvascular endothelial growth factor | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human/Cynomolgus VEGF 165 (Uniprot#: P15692-4; Met 1-Arg 191). | |
0.1 mL | |
Angiogenesis, Biologically Active Proteins | |
7422 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction