Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VEGF 165 Antibody (Bevacizumab), DyLight 650, Novus Biologicals™

Human Monoclonal Antibody
Supplier: Novus Biologicals NBP259637C
Description
VEGF 165 Monoclonal specifically detects VEGF 165 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
VEGF 165 | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate | |
Human | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, ELISA | |
Bevacizumab | |
Western Blot, Flow Cytometry, ELISA | |
MVCD1, vascular endothelial growth factor A, Vascular permeability factor, VEGF-A, VEGFMGC70609, VPFvascular endothelial growth factor | |
Recombinant human VEGF | |
0.1 mL | |
Angiogenesis, Biologically Active Proteins | |
7422 | |
Store at 4°C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction