Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human VEGF-165 Recombinant Protein

Catalog No. ENRVEGFI
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ENRVEGFI 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ENRVEGFI Supplier Invitrogen™ Supplier No. RVEGFI
Only null left
Add to Cart
Edge
Add to Cart

Recombinant Protein

Mature human VEGF165 was expressed in Pichia pastoris, KM71. GenBank Accession # 003376, amino acids 27 - 191. For protein verification, the expressed protein was N-terminal sequenced and begins with the following 10 amino acids: EGGGQNHHEV. Purity is >95% by SDS-PAGE analyses. Activity: The biological activity of recombinant human VEGF165 was determined by measuring the proliferation of human umbilical vein endothelial cells (HUVEC). Use: For in vitro applications. A dose-response assay should be performed to determine the optimal concentration for specific experiments. Reconstitution and Dilution: Store lyophilized material at 4°C. Reconstitute in 1 mL of distilled or deionized water. Dilute in a buffer-containing carrier protein, aliquot, and store at or below -70°C, avoiding multiple freeze-thaw cycles. PBS with 1% BSA or a neutral media containing <10% FCS are suitable buffers.

The vascular endothelial growth factor (VEGF) family currently includes VEGF-A, VEGF-B, VEGF-C, VEGF-D, VEGF-E, and PIGF. VEGF and its receptor system have been shown to be the fundamental regulators in the cell signaling of angiogenesis. Most tumors have the absolute requirement of angiogenesis and VEGF has been described as the most potent angiogenic cytokine linked to this process. To date 5 different isoforms of VEGF have been described. These isoforms are generated as the result of alternative splicing from a single VEGF gene. These various isoforms have been shown to bind to two tyrosine-kinase receptors flt-1 (VEGFR-1) and flk-1/KDR (VEGFR-2), which have been found to be expressed almost exclusively on endothelial cells.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P15692
For Use With (Application) Bioactivity
Formulation PBS with no preservative
Gene ID (Entrez) 7422
Molecular Weight (g/mol) 19.2 kDa
Name Human VEGF-165
Purification Method Purified
Quantity 20 μg
Source Yeast
Storage Requirements 4°C
Regulatory Status RUO
Gene Alias H-VEGF-165; MVCD1; M-VEGF-165; vascular endothelial growth factor 165b; vascular endothelial growth factor A; vascular endothelial growth factor A121; vascular endothelial growth factor A165; vascular permeability factor; VEGF; VEGF164; VEGF165; Vegfa; VEGF-A; VPF
Common Name VEGF-165
Gene Symbol Vegfa
Biological Activity Determined by the proliferation effect on human umbilical vein endothelial cells (HUVECs).
Product Type Protein
Conjugate Unconjugated
Species Yeast
Recombinant Recombinant
Sequence Human VEGF-165, amino acids 27-191 (Accession # 003376)
Content And Storage 4°C
Activity Determined by the proliferation effect on human umbilical vein endothelial cells (HUVECs).
Expression System P. pastoris
Form Lyophilized
Protein Subtype Recombinant
Purity or Quality Grade >95% by SDS-PAGE
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.