Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VEGF Antibody, Alexa Fluor™ 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB1002381AF350
Description
VEGF Polyclonal specifically detects VEGF in Human, Mouse, Rat, Bovine, Canine, Chicken, Equine, Guinea Pig samples. It is validated for Western Blot, ELISA.Specifications
VEGF | |
Polyclonal | |
Western Blot, ELISA | |
MVCD1, vascular endothelial growth factor A, Vascular permeability factor, VEGF-A, VEGFMGC70609, VPFvascular endothelial growth factor | |
Rabbit | |
Affinity Purified | |
Angiogenesis, Cancer, Cell Biology, Cellular Markers, GPCR, HIF Target Genes, Hypoxia, Metastasis, Neuroscience, Neurotransmission, Sensory Systems, Signal Transduction, Vision | |
7422.0 | |
Store at 4°C in the dark. |
Western Blot, ELISA | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
VEGFA | |
A synthetic peptide made to an internal region of the human VEGFA protein sequence (between residues 180-232). [Swiss-Prot: P15692] | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Bovine, Canine, Chicken, Equine, Guinea Pig | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction