Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VEGF Antibody, Alexa Fluor™ 647, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB1002381AF647
Description
VEGF Polyclonal specifically detects VEGF in Human, Mouse, Rat, Bovine, Canine, Chicken, Equine, Guinea Pig samples. It is validated for Western Blot, ELISA.Specifications
VEGF | |
Polyclonal | |
Western Blot, ELISA | |
P15692 | |
VEGFA | |
A synthetic peptide made to an internal region of the human VEGFA protein sequence (between residues 180-232). [Swiss-Prot: P15692] | |
0.1 mL | |
Angiogenesis, Cancer, Cell Biology, Cellular Markers, GPCR, HIF Target Genes, Hypoxia, Neuroscience, Neurotransmission, Sensory Systems, Signal Transduction, Vision | |
7422 | |
Human, Mouse, Rat, Bovine, Canine, Chicken, Equine, Guinea Pig | |
IgG |
Western Blot, ELISA | |
Alexa Fluor 647 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
MVCD1, vascular endothelial growth factor A, Vascular permeability factor, VEGF-A, VEGFMGC70609, VPFvascular endothelial growth factor | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human, mouse, rat, horse, chicken, guinea pig, dog, and cow. Predicted to react with sheep and primate based on 100% sequence homology. | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction