Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VEGF Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 5 publications
$206.50 - $499.50
Specifications
Antigen | VEGF |
---|---|
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1 ug/ml, ELISA 1:500 |
Applications | Western Blot, ELISA |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB1002381SS
![]() |
Novus Biologicals
NB1002381SS |
0.025 mL |
Each for $206.50
|
|
|||||
NB1002381
![]() |
Novus Biologicals
NB1002381 |
0.1 mL |
Each for $499.50
|
|
|||||
Description
VEGF Polyclonal specifically detects VEGF in Human, Mouse, Rat, Bovine, Canine, Chicken, Equine, Guinea Pig samples. It is validated for Western Blot, ELISA.Specifications
VEGF | |
Western Blot 1 ug/ml, ELISA 1:500 | |
Polyclonal | |
Rabbit | |
Angiogenesis, Cancer, Cell Biology, HIF Target Genes, Hypoxia, Signal Transduction | |
PBS with 0.09% Sodium Azide | |
MVCD1, vascular endothelial growth factor A, Vascular permeability factor, VEGF-A, VEGFMGC70609, VPFvascular endothelial growth factor | |
VEGFA | |
IgG | |
Affinity Purified | |
Human, mouse, rat, horse, chicken, guinea pig, dog, and cow. Predicted to react with sheep and primate based on 100% sequence homology. |
1.0 mg/mL | |
Western Blot, ELISA | |
Unconjugated | |
RUO | |
Human, Mouse, Rat, Bovine, Canine, Chicken, Equine, Guinea Pig | |
P15692 | |
7422 | |
A synthetic peptide made to an internal region of the human VEGFA protein sequence (between residues 180-232). [Swiss-Prot: P15692] | |
Primary | |
Store at 4C. Do not freeze. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title