Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VEGF Antibody (VG1) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 61 publication
$244.50 - $574.50
Specifications
Antigen | VEGF |
---|---|
Clone | VG1 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1-2 ug/ml, Simple Western, Flow Cytometry, ELISA, Immunohistochemistry 1:20-1:100, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:20-1:100, Immunohistochemistry-Frozen 1:20-1:100, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), CyTOF |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB100664SS
![]() |
Novus Biologicals
NB100664SS |
0.025 mg |
Each for $244.50
|
|
|||||
NB100664
![]() |
Novus Biologicals
NB100664 |
0.1 mg |
Each for $574.50
|
|
|||||
Description
VEGF Monoclonal specifically detects VEGF in Human, Mouse, Rat, Porcine, Canine samples. It is validated for Western Blot, Simple Western, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, CyTOF-ready.Specifications
VEGF | |
1 mg/mL | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), CyTOF | |
Unconjugated | |
Mouse | |
Angiogenesis, Cancer, Cell Biology, HIF Target Genes, Hypoxia, Metastasis | |
P15692 | |
7422 | |
Recombinant VEGF 189 protein. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
VG1 | |
Western Blot 1-2 ug/ml, Simple Western, Flow Cytometry, ELISA, Immunohistochemistry 1:20-1:100, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:20-1:100, Immunohistochemistry-Frozen 1:20-1:100, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat, Pig, Canine | |
MVCD1, vascular endothelial growth factor A, Vascular permeability factor, VEGF-A, VEGFMGC70609, VPFvascular endothelial growth factor | |
VEGFA | |
IgG1 κ | |
Protein G purified | |
This VEGF Antibody (VG1) detects the 189, 165 and 121 isoforms of VEGF |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title