Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VEGF Antibody (VG76e) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP281006
Description
VEGF Monoclonal specifically detects VEGF in Human, Rat, Porcine, Bovine, Sheep samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Radioimmunoassay.Specifications
VEGF | |
Monoclonal | |
1 mg/mL | |
Western Blot : 0.5-2ug/ml, ELISA : 1:100-1:2000, Immunohistochemistry : 1:200, Immunocytochemistry/Immunofluorescence : 1:50-1:200, Immunohistochemistry-Paraffin 1:200, Radioimmunoassay reported in scientific literature | |
MVCD1, vascular endothelial growth factor A, Vascular permeability factor, VEGF-A, VEGFMGC70609, VPFvascular endothelial growth factor | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human. Mixed results in Cow and Pig (IHC only). Customers have reported success staining sheep tissue (see review). | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Radioimmune Assays (RIA) | |
VG76e | |
Unconjugated | |
PBS | |
VEGFA | |
Recombinant human VEGF189 expressed in E.coli [UniProt# P15692] | |
0.1 mL | |
Angiogenesis, Cancer, Cell Biology, Cellular Markers, GPCR, HIF Target Genes, Hypoxia, Metastasis, Neuroscience, Neurotransmission, Sensory Systems, Signal Transduction, Vision | |
7422 | |
Human, Rat, Pig, Bovine, Sheep | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction