Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VEGF Antibody (VG76e), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB100648F
Description
VEGF Monoclonal specifically detects VEGF in Human, Rat, Porcine, Bovine, Sheep samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
VEGF | |
Monoclonal | |
FITC | |
P15692 | |
VEGFA | |
Recombinant human VEGF189 expressed in E.coli [UniProt# P15692] | |
Protein G purified | |
RUO | |
Primary | |
Using recombinant proteins, it was demonstrated that VG76e recognizes the 121, 165, and 189 isoforms of VEGF | |
Store at 4C in the dark. | |
IgG1 |
ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
VG76e | |
ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
MVCD1, vascular endothelial growth factor A, Vascular permeability factor, VEGF-A, VEGFMGC70609, VPFvascular endothelial growth factor | |
Mouse | |
40 kDa | |
0.1 mL | |
Angiogenesis, Cancer, Cell Biology, HIF Target Genes, Hypoxia | |
7422 | |
Human, Rat, Pig, Bovine, Sheep | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction