Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VEGF-B Antibody (OTI1H9), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274844G
Description
VEGF-B Monoclonal specifically detects VEGF-B in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
VEGF-B | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
VEGFB | |
Full length recombinant protein of human VEGF-B (NP_003368) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI1H9 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
vascular endothelial growth factor B, VEGF-B, VEGF-related factor, VRFVEGFL | |
Mouse | |
Affinity Purified | |
Angiogenesis, Cell Cycle and Replication | |
7423.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction