Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VEGF-B Antibody (RM0008-6E72), mFluor Violet 610 SE, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NB11061024MFV610
Description
VEGF-B Monoclonal antibody specifically detects VEGF-B in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
VEGF-B | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rat | |
Protein G purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
RM0008-6E72 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
vascular endothelial growth factor B, VEGF-B, VEGF-related factor, VRFVEGFL | |
Mouse recombinant VEGF-B | |
0.1 mL | |
Angiogenesis, Cell Cycle and Replication | |
7423 | |
Store at 4C in the dark. | |
IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction