Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VEGF-B Antibody (RM0008-6E72), PerCP, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NB11061024PCP
Description
VEGF-B Monoclonal antibody specifically detects VEGF-B in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| VEGF-B | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| RM0008-6E72 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| vascular endothelial growth factor B, VEGF-B, VEGF-related factor, VRFVEGFL | |
| Mouse recombinant VEGF-B | |
| 0.1 mL | |
| Angiogenesis, Cell Cycle and Replication | |
| 7423 | |
| Store at 4C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction