Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VEGF-B Antibody (RM0008-6E72), Alexa Fluor™ 750, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NB11061024AF750
Description
VEGF-B Monoclonal specifically detects VEGF-B in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
VEGF-B | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
VEGFB | |
Mouse recombinant VEGF-B | |
0.1 mL | |
Primary | |
This antibody recognizes mouse VEGF-B 186 but not 167 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
RM0008-6E72 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
vascular endothelial growth factor B, VEGF-B, VEGF-related factor, VRFVEGFL | |
Rat | |
Protein G purified | |
Angiogenesis, Cell Cycle and Replication | |
7423.0 | |
Mouse | |
IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction