Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VEGF-C Antibody (107/A11), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP2362880.02MG
Description
VEGF-C Monoclonal specifically detects VEGF-C in Human, Rat samples. It is validated for Western Blot, ELISA, Sandwich ELISA.Specifications
VEGF-C | |
Monoclonal | |
LYOPH | |
Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Sandwich ELISA 1:100-1:2000 | |
Flt4 ligand, Flt4-L, vascular endothelial growth factor C, Vascular endothelial growth factor-related protein, VEGF-C, VRPFLT4 ligand DHM | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Centrifuge vial prior to opening. Reconstitute in sterile water to a concentration of 0.1-1.0 mg/ml. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG2a |
ELISA, Western Blot | |
107/A11 | |
Unconjugated | |
PBS and 50x BSA with 0.02% Sodium Azide | |
VEGFC | |
Recombinant human VEGF-C | |
0.02 mg | |
Angiogenesis, Cancer, Cell Biology, HIF Target Genes, Hypoxia | |
7424 | |
Rat, Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction