Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VEGF-C Antibody (9/G10), Alexa Fluor™ 350, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP236287AF350

Catalog No. NP236287A30


Only null left
Add to Cart

Description

Description

VEGF-C Monoclonal specifically detects VEGF-C in Human, Rat samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

VEGF-C
Monoclonal
Alexa Fluor 350
50 mM sodium borate with 0.05% sodium azide
VEGFC
Recombinant human VEGF-C
0.1 mL
Primary
Human, Rat
IgG2a
Western Blot, ELISA
9/G10
Western Blot, ELISA
Flt4 ligand, Flt4-L, vascular endothelial growth factor C, Vascular endothelial growth factor-related protein, VEGF-C, VRPFLT4 ligand DHM
Mouse
Protein G purified
Angiogenesis, Cancer, Cell Biology, HIF Target Genes, Hypoxia
7424.0
Store at 4°C in the dark.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.