Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VEGF-C Antibody (9/G10) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$243.50 - $560.50
Specifications
| Antigen | VEGF-C |
|---|---|
| Clone | 9/G10 |
| Concentration | LYOPH |
| Dilution | Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Sandwich ELISA Capture 1:100-1:2000 |
| Applications | ELISA, Western Blot |
Description
VEGF-C Monoclonal specifically detects VEGF-C in Human, Rat samples. It is validated for Western Blot, ELISA, Sandwich ELISA Capture.Specifications
| VEGF-C | |
| LYOPH | |
| ELISA, Western Blot | |
| Unconjugated | |
| Mouse | |
| Angiogenesis, Cancer, Cell Biology, HIF Target Genes, Hypoxia | |
| Flt4 ligand, Flt4-L, vascular endothelial growth factor C, Vascular endothelial growth factor-related protein, VEGF-C, VRPFLT4 ligand DHM | |
| VEGFC | |
| IgG2a | |
| Protein G purified | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 9/G10 | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Sandwich ELISA Capture 1:100-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Rat | |
| 7424 | |
| Recombinant human VEGF-C | |
| Primary | |
| Centrifuge vial prior to opening. Reconstitute in sterile water to a concentration of 0.1-1.0 mg/ml. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title