Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VEGF-C Antibody (OTI4A1), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274845IR
Description
VEGF-C Monoclonal specifically detects VEGF-C in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
VEGF-C | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
VEGFC | |
Recombinant protein fragment corresponding to amino acids 112-227 of human VEGF-C (NP_005420) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot | |
OTI4A1 | |
Western Blot | |
Flt4 ligand, Flt4-L, vascular endothelial growth factor C, Vascular endothelial growth factor-related protein, VEGF-C, VRPFLT4 ligand DHM | |
Mouse | |
Affinity Purified | |
Angiogenesis, Cancer, Cell Biology, HIF Target Genes, Hypoxia | |
7424.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction