Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VEGF-D Antibody (106), PE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289563PE
Description
VEGF-D Monoclonal antibody specifically detects VEGF-D in Human samples. It is validated for ELISASpecifications
VEGF-D | |
Monoclonal | |
PE | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
106 | |
ELISA | |
c-fos induced growth factor (vascular endothelial growth factor D), vascular endothelial growth factor D, VEGF-DVEGFDc-Fos-induced growth factor | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human VEGF-D (Accession#: NP_004460.1; Phe 93-Ser 201) . | |
0.1 mL | |
Angiogenesis, Cancer, Cellular Markers | |
2277 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction