Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VEGF-D Antibody (2-2#), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP341061
Description
VEGF-D Monoclonal antibody specifically detects VEGF-D in Human,Rat samples. It is validated for ELISA,Western Blot,Immunohistochemistry (Paraffin),Immunohistochemistry (Frozen),Immunocytochemistry/ ImmunofluorescenceSpecifications
VEGF-D | |
Monoclonal | |
1 mg/mL | |
Western Blot 0.5-5ug/mL, ELISA 0.05-2ug/mL, Immunocytochemistry/ Immunofluorescence 5-30ug/mL, Immunohistochemistry-Paraffin 5-30ug/mL, Immunohistochemistry-Frozen 5-30ug/mL | |
c-fos induced growth factor (vascular endothelial growth factor D), vascular endothelial growth factor D, VEGF-DVEGFDc-Fos-induced growth factor | |
Immunized with recombinant Human VEGF-D (Uniprot#: O43915; Phe93-Ser201 (Uniprot#: O43915) | |
100 μL | |
Angiogenesis, Cancer, Cellular Markers | |
2277 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG2b κ |
ELISA, Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunocytochemistry/Immunofluorescence | |
2-2# | |
Unconjugated | |
PBS, pH 7.4, 50% glycerol | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction