Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VEGF-D Antibody (MM0007-7E79), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB11060973IR
Description
VEGF-D Monoclonal specifically detects VEGF-D in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
VEGF-D | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
VEGFD | |
Human recombinant VEGF-D | |
0.1 mL | |
Primary | |
VEGFD (MM0007-7E79). No cross-reactivity is shown with VEGF and VEGF-B. | |
Store at 4C in the dark. | |
IgG2 |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
MM0007-7E79 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
c-fos induced growth factor (vascular endothelial growth factor D), vascular endothelial growth factor D, VEGF-DVEGFDc-Fos-induced growth factor | |
Mouse | |
Protein G purified | |
Angiogenesis, Cancer, Cellular Markers | |
2277 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction