Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VEGF-164 Monoclonal Antibody (123704)
GREENER_CHOICE

Catalog No. PIMA523769
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA523769 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA523769 Supplier Invitrogen™ Supplier No. MA523769
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody shows 50-100% cross-reactivity with recombinant mouse (rm) VEGF120, rmVEGF164, recombinant human (rh) VEGF121, rhVEGF165, and rhVEGF189 and no cross-reactivity with rmVEGF115. This antibody recognizes an epitope found between amino acids 78-120 of rmVEGF120. This segment is present in all VEGF isoforms except rmVEGF115. Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.

VEGF-164 is a gene target that encodes for the vascular endothelial growth factor. This protein is responsible for promoting angiogenesis. VEGF-164 has been shown to be involved in the development of various diseases, including cancer, cardiovascular disease, and diabetic retinopathy. Targeting this gene has become an important strategy in the development of new therapeutics for these conditions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VEGF-164
Applications ELISA, Western Blot
Classification Monoclonal
Clone 123704
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene VEGFA
Gene Accession No. P16612
Gene Alias Vascular Endothelial Growth Factor, Vascular Permeability Factor, VEGFA, VPF
Gene Symbols VEGFA
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant rat VEGF164 Ala27-Arg190.
Purification Method Protein A/G
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83785
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.