Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ VEGF, Rabbit, Alexa Fluor 488, Polyclonal,

Catalog No. ABS82AF488 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Alexa Fluor 488
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ABS82AF488 100 μL Alexa Fluor 488
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS82AF488 Supplier MilliporeSigma™ Supplier No. ABS82AF488
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Keratin, type I cytoskeletal 18 (UniProt P05783; also known as Cell proliferation-inducing gene 46 protein, CK-18, Cytokeratin-18, K18, Keratin-18) is encoded by the KRT18 (also known as CYK18, PIG46) gene (Gene ID 3875) in human. Cytokeratin-18 (Keratin-18 or K18) belongs to a family of highly conserved epithelial-specific intermediate filament (IF) proteins that play an important role in the mechanical stability and integrity of epithelial cells and tissues. Each epithelium is characterized by its unique keratin expression pattern. All epithelial cells typically express at least one type-I and one type-II keratin, which form noncovalent heteropolymers. Keratin 18 is a type I intermediate filament protein that pairs with keratin 8 (K8/K18) in simple epithelia and plays an important role in protecting cells from stress, injury, and apoptosis. K18 is the only type-I keratin in adult hepatocytes and the major type-I keratin in many colonic tumor cell lines.The structure and function of K8/18 are regulated by posttranslational modifications, such as glycosylation, ubiquitination, and phosphorylation, including K8 phosphorylation on Ser8, Ser23, Ser73, Ser431, and K18 phosphorylation on Ser33 and Ser52. K18 phosphorylation has been proposed as a progression marker for human chronic hepatitis B (CHB). Progressively higher Ser52 phosphorylation is reported in non-cirrhotic and cirrhotic CHB livers, and progressive increase of Ser33 and Ser52 phosphorylation correlates with the elevation of both histological lesions and enzymatic activities of alanine aminotransferase in non-cirrhotic CHB livers.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VEGF
Applications Immunocytochemistry
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Alexa Fluor 488
Formulation Purified rabbit polyclonal antibody conjugate in PBS with 15mg/ml BSA and 0.1 % Sodium Azide.
Gene VEGFA, VEGF
Gene Accession No. P15692
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to human VEGF at the N-terminus.
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_001020537
Test Specificity This antibody recognizes VEGF at the N-terminus.
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.