Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VEGF Receptor 1/2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. APPA6612
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
APPA6612 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. APPA6612 Supplier Invitrogen™ Supplier No. PA116612
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The target sequence has 100% sequence homology with mouse and rat.

Receptors for VEGF, VEGFB and PGF have a tyrosine-protein kinase activity. The VEGF-kinase ligand/receptor signaling system plays a key role in vascular development and regulation of vascular permeability. VEGF and its high-affinity binding receptors, the tyrosine kinases FLK1 and FLT1, are thought to be important for the development of embryonic vasculature. It has been shown that an alternately spliced form of FLT1 produces a soluble protein, termed sFLT1, which binds vascular endothelial growth factor with high affinity, playing an inhibitory role in angiogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VEGF Receptor 1/2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene FLT1
Gene Accession No. O08775, P17948, P35968, P53767
Gene Alias 6130401C07; AI323757; CD309; Embryonic receptor kinase 2; Emrk2; Fetal liver kinase 1; fetal liver kinase-1; Flk1; FLK-1; FLK1 kinase insert domain receptor (a type III receptor tyrosine kinase) (VEGF receptor 2); FLK1 kinase insert domain receptor (VEGF receptor 2); FLT; Flt1; FLT-1; fms related tyrosine kinase 1; FMS-like tyrosine kinase 1; FMS-related tyrosine kinase 1; Fms-related tyrosine kinase 1 (vascular endothelial growth factor/vascular permeability factor receptor); FRT; KDR; kinase insert domain protein receptor; kinase insert domain receptor; kinase insert domain receptor (a type III receptor tyrosine kinase); kinase NYK; Krd-1; Ly73; Protein-tyrosine kinase receptor flk-1; sFlt1; soluble vascular endothelial growth factor receptor 2; soluble VEGFR 1; soluble VEGFR2; sVEGFR 1; sVEGFR-2; tyrosine kinase growth factor receptor; tyrosine-protein kinase FRT; Tyrosine-protein kinase receptor FLT; vascular endothelial growth factor receptor 1; Vascular endothelial growth factor receptor 2; vascular endothelial growth factor receptor- 2; vascular endothelial growth factor receptor-1; vascular endothelial growth factor receptor-2; vascular endothelial growth factor receptor-3; vascular permeability factor receptor; VEGF R1; VEGF receptor-2; VEGFR; VEGFR 1; Vegfr1; VEGFR-1; VEGFR2; Vegfr-2
Gene Symbols FLT1, KDR
Host Species Rabbit
Immunogen A synthetic peptide made to an internal region of the mouse VEGF Receptor 1 protein (between residues 800-900).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2321, 25589, 3791, 54251
Target Species Human, Rat
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.