Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VEGFA Mouse anti-Human, Clone: VEGF/1063, Abnova™

Catalog No. 89034730 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-034-730 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-034-730 Supplier Abnova Corporation Supplier No. MAB14647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against full length recombinant human VEGFA.

  • High specificity
  • High reactivity

Specifications

Antigen VEGFA
Applications Immunofluorescence, Immunohistochemistry (PFA fixed)
Classification Monoclonal
Clone VEGF/1063
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant human VEGFA.
Dilution Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (2-4 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS.
Gene Accession No. P15692-2
Gene Alias MGC70609/VEGF/VEGF-A/VPF
Gene Symbols VEGFA
Host Species Mouse
Immunogen Recombinant protein corresponding to full length human VEGFA isoform VEGFA189.
Purification Method Protein G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7422
Test Specificity This antibody is highly specific to VEGFA and recognizes the 19-22kDa (reducing) and 38-44kDa (non-reducing) form of VEGFA.
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.