Learn More
Invitrogen™ VEGFB Monoclonal Antibody (OTI1H9)

Description
VEGFB Monoclonal Antibody for Western Blot, IHC (P), ICC/IF
Specifications
Specifications
| Antigen | VEGFB |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | OTI1H9 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | VEGFB |
| Gene Accession No. | P49765 |
| Gene Alias | flt-1/VEGFR1 ligand; vascular endothelial growth factor B; vascular endothelial growth factor B167 precursor; vascular endothelial growth factor B186 precursor; vascular endothelial growth factor-B; Vegfb; VEGF-B; VEGFL; VEGF-related factor; Vrf |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.