Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VEGFC Monoclonal Antibody (30AT1174.77)
GREENER_CHOICE

Catalog No. PIMA511119
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA511119 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA511119 Supplier Invitrogen™ Supplier No. MA511119
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

VEGFC Monoclonal Antibody for Western Blot

The protein encoded by this gene is a member of the platelet-derived growth factor/vascular endothelial growth factor (PDGF/VEGF) family, is active in angiogenesis and endothelial cell growth, and can also affect the permeability of blood vessels. This secreted protein undergoes a complex proteolytic maturation, generating multiple processed forms which bind and activate VEGFR-3 receptors. Only the fully processed form can bind and activate VEGFR-2 receptors. This protein is structurally and functionally similar to vascular endothelial growth factor D.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VEGFC
Applications Western Blot
Classification Monoclonal
Clone 30AT1174.77
Concentration 0.45 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene VEGFC
Gene Accession No. P49767, P97953
Gene Alias AW228853; Flt4 ligand; FLT4 ligand DHM; Flt4-L; LMPH1D; Vascular endothelial growth factor C; vascular endothelial growth factor C isoform 129; vascular endothelial growth factor C isoform 184; vascular endothelial growth factor-related protein; VEGF3; VEGFC; VEGF-C; VRP
Gene Symbols VEGFC
Host Species Mouse
Immunogen KLH conjugated synthetic peptides selected from the N-terminal, central, and C-terminal regions of human VEGF3.
Purification Method Protein G
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22341, 7424
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.