Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VEGFC Propeptide Polyclonal Antibody, Invitrogen™

Catalog No. 344300
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-430-0 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 34-430-0 Supplier Invitrogen Supplier No. 344300
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

VEGFC Propeptide Polyclonal antibody specifically detects VEGFC Propeptide in Human samples. It is validated for ELISA, Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VEGFC Propeptide
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene VEGFC
Gene Accession No. P49767
Gene Alias Flt4 ligand; FLT4 ligand DHM; Flt4-L; LMPH1D; vascular endothelial growth factor C; Vascular endothelial growth factor-related protein; VEGF3; VEGFC; VEGF-C; VRP
Gene Symbols VEGFC
Host Species Rabbit
Immunogen A synthetic peptide derived from the C-terminal sequence of propeptide form of VEGF-C.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7424
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.