Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VEGFD Polyclonal Antibody
VEGFD Polyclonal Antibody
Supplier: Thermo Scientific PA594961
Description
VEGFD Polyclonal Antibody for Western Blot, IHC (P)

Specifications
VEGFD | |
Polyclonal | |
Unconjugated | |
VEGFD | |
AI325264; c-fos induced growth factor; c-fos induced growth factor (vascular endothelial growth factor D); c-fos-induced growth factor; FIGF; Vascular endothelial growth factor D; vascular endothelial growth factor-D; VEGF4; VEGFD; VEGF-D; VEGFR2 and -3 ligand | |
Rabbit | |
Affinity chromatography | |
RUO | |
14205, 2277, 360457 | |
-20°C | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
500 μg/mL | |
PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal | |
O35251, O43915, P97946 | |
VEGFD | |
A synthetic peptide corresponding to a sequence in the middle region of human VEGFD (101-115aa VIDEEWQRTQCSPRE). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction