Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ VEGFD Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA579266
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: rat lung tissue, rat brain tissue, mouse lung tissue, mouse brain tissue. IHC: human colon cancer tissue, human lung cancer tissue, human lung cancer tissue.
The protein encoded by this gene is a member of the platelet-derived growth factor/vascular endothelial growth factor (PDGF/VEGF) family and is active in angiogenesis, lymphangiogenesis, and endothelial cell growth. This secreted protein undergoes a complex proteolytic maturation, generating multiple processed forms which bind and activate VEGFR-2 and VEGFR-3 receptors. This protein is structurally and functionally similar to vascular endothelial growth factor C.
Specifications
VEGFD | |
Polyclonal | |
Unconjugated | |
VEGFD | |
AI325264; c-fos induced growth factor; c-fos induced growth factor (vascular endothelial growth factor D); c-fos-induced growth factor; FIGF; Vascular endothelial growth factor D; vascular endothelial growth factor-D; VEGF4; VEGFD; VEGF-D; VEGFR2 and -3 ligand | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14205, 2277, 360457 | |
-20°C | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
500 μg/mL | |
PBS with 5mg BSA and 0.05mg sodium azide | |
O35251, O43915, P97946 | |
VEGFD | |
E. coli-derived human VEGFD recombinant protein (Position: F89-R205). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction