Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VEGFR1/Flt-1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NB100685
Description
VEGFR1/Flt-1 Polyclonal specifically detects VEGFR1/Flt-1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| VEGFR1/Flt-1 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.4), 0.2% BSA, Tween-20 with 0.05% Sodium Azide | |
| EC 2.7.10, EC 2.7.10.1, FLT, FLT-1, Fms-like tyrosine kinase 1, fms-related tyrosine kinase 1 (vascular endothelial growth factor/vascularpermeability factor receptor), FRT, Tyrosine-protein kinase FRT, Tyrosine-protein kinase receptor FLT, vascular endothelial growth factor receptor 1, Vascular permeability factor receptor, VEGFR1, VEGFR-1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This reacts with a 180 kD protein, known as VEGFR1/Flt-1 or vascular endothelial growth factor receptor 1. VEGFR1/Flt-1 is a cell membrane receptor kinase. VEGFR1/Flt-1 has a high affinity receptor for vascular endothelial growth factor and is putatively involved in the growth of endothelial cells and angiogenesis. | |
| Store at 4C. Do not freeze. |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| 0.5 mg/mL | |
| Immunohistochemistry 1:25-1:50, Immunohistochemistry-Paraffin 1:25-1:50, Immunohistochemistry-Frozen 1:50-1:100 | |
| P17948 | |
| FLT1 | |
| Synthetic peptide: C-AELERKIACCSPPPDYN, corresponding to amino acids 1312-1328 of human VEGF Receptor 1 (C-terminus). | |
| 0.5 mL | |
| Angiogenesis, Cancer, HIF Target Genes, Hypoxia, Stem Cell Markers | |
| 2321 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction