Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VEGFR1/Flt-1 Antibody (FLT1/658), Alexa Fluor™ 532, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP234618AF532

Catalog No. NP234618A32


Only null left
Add to Cart

Description

Description

VEGFR1/Flt-1 Monoclonal specifically detects VEGFR1/Flt-1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

VEGFR1/Flt-1
Monoclonal
Alexa Fluor 532
50 mM sodium borate with 0.05% sodium azide
FLT1
Recombinant human VEGFR1/Flt-1 protein (Uniprot: P17948)
0.1 mL
Primary
Three cell membrane receptor tyrosine kinases, Flt-1 (also designated VEGF-R1), Flk-1 (also designated VEGF-R2) and Flt-4, putatively involved in the growth of endothelial cells, are characterized by the presence of seven immunoglobulin-like sequences in their extracellular domain. These receptors exhibit high degrees of sequence relatedness to each other as well as lesser degrees of relatedness to the class III receptors including CSF-1/Fms, PDGR, SLFR/Kit and Flt-3/Flk-2. Two members of this receptor class, Flt-1 and Flk-1, have been shown to represent high affinity receptors for vascular endothelial growth factors (VEGFs). On the basis of structural similarity to Flt-1 and Flk-1, it has been speculated that Flt-4 might represent a third receptor for either VEGF or a VEGF-related ligand.
Store at 4°C in the dark.
ELISA
FLT1/658
ELISA
EC 2.7.10, EC 2.7.10.1, FLT, FLT-1, Fms-like tyrosine kinase 1, fms-related tyrosine kinase 1 (vascular endothelial growth factor/vascularpermeability factor receptor), FRT, Tyrosine-protein kinase FRT, Tyrosine-protein kinase receptor FLT, vascular endothelial growth factor receptor 1, Vascular permeability factor receptor, VEGFR1, VEGFR-1
Mouse
Protein A or G purified
Angiogenesis, Cancer, Cellular Markers, Endothelial Cell Markers, HIF Target Genes, Hypoxia, Myeloid derived Suppressor Cell, Phospho Specific, Signal Transduction, Stem Cell Markers, Tumor Suppressors, Tyrosine Kinases
2321
Human
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.