Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VEGFR1/R2/R3 Antibody (FLT1/1663), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314285MFV450
Description
VEGFR1/R2/R3 Monoclonal antibody specifically detects VEGFR1/R2/R3 in Human samples. It is validated for Flow CytometrySpecifications
VEGFR1/R2/R3 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
2321 | |
Store at 4C in the dark. | |
IgG1 κ |
Flow Cytometry | |
FLT1/1663 | |
Flow Cytometry | |
FLT;FLT1;FLT-1;Receptor protein-tyrosine kinase;Vascular endothelial growth factor receptor 1;VEGFR1;VEGFR-1 | |
Recombinant human VEGFR1/R2/R3 protein (Uniprot: P17948) | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction