Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VEGFR1/R2/R3 Mouse anti-Human, Clone: FLT1/1663, Novus Biologicals™

Mouse Monoclonal Antibody
$240.00 - $492.50
Specifications
Antigen | VEGFR1/R2/R3 |
---|---|
Clone | FLT1/1663 |
Concentration | 0.2 mg/ml |
Dilution | Flow Cytometry 1-2 ug/million cells, Immunofluorescence 1-2 ug/ml |
Applications | Flow Cytometry, Immunofluorescence |
Description
VEGFR1/R2/R3 Monoclonal antibody specifically detects VEGFR1/R2/R3 in Human samples. It is validated for Flow Cytometry, ImmunofluorescenceSpecifications
VEGFR1/R2/R3 | |
0.2 mg/ml | |
Flow Cytometry, Immunofluorescence | |
Unconjugated | |
Mouse | |
Human | |
P17948 | |
Recombinant human VEGFR1/R2/R3 protein (Uniprot: P17948) | |
Primary | |
Store at 4°C. |
FLT1/1663 | |
Flow Cytometry 1-2 ug/million cells, Immunofluorescence 1-2 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
10 mM PBS with 0.05% BSA | |
2321 | |
IgG1 κ | |
Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title