Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VEGFR1 Rabbit anti-Human, Non-human Primate, Polyclonal, FabGennix

Catalog No. 502209602
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-9602 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-9602 Supplier Fabgennix Inc Supplier No. VEGFR1101AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

VEGFR1 Polyclonal antibody specifically detects VEGFR1 in Human, Non-human Primate samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot

Specifications

Antigen VEGFR1
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene FLT1
Gene Accession No. P17948
Gene Alias AI323757; embryonic receptor kinase 2; Emrk2; FLT; FLT1; FLT-1; fms related tyrosine kinase 1; FMS-like tyrosine kinase 1; FMS-related tyrosine kinase 1; Fms-related tyrosine kinase 1 (vascular endothelial growth factor/vascular permeability factor receptor); FRT; i21; sFlt1; soluble VEGFR 1; sVEGFR 1; Tyrosine-protein kinase FRT; tyrosine-protein kinase receptor FLT; Vascular endothelial growth factor receptor 1; vascular endothelial growth factor receptor-1; vascular permeability factor receptor; VEGF R1; VEGFR1; VEGFR-1
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 1250-1300 on human Vascular endothelial growth factor receptor 1 protein.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2321, 452512
Target Species Human, Non-human Primate
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.