Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VEGFR2/KDR/Flk-1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 4 publications
$192.47 - $529.95
Specifications
| Antigen | VEGFR2/KDR/Flk-1 |
|---|---|
| Concentration | 2.55 mg/mL |
| Dilution | Western Blot 1:250-1:1000, Flow Cytometry reported in scientific literature (PMID 29873146), Immunohistochemistry reported in scientific literature (PMID 30941660), Immunocytochemistry/Immunofluorescence 1:50 - 1:500 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB100530SS
![]() |
Novus Biologicals
NB100530SS |
0.025 mL |
Each for $192.47
|
|
|||||
NB100530
![]() |
Novus Biologicals
NB100530 |
0.1 mL |
Each for $529.95
|
|
|||||
Description
VEGFR2/KDR/Flk-1 Polyclonal specifically detects VEGFR2/KDR/Flk-1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
| VEGFR2/KDR/Flk-1 | |
| Western Blot 1:250-1:1000, Flow Cytometry reported in scientific literature (PMID 29873146), Immunohistochemistry reported in scientific literature (PMID 30941660), Immunocytochemistry/Immunofluorescence 1:50 - 1:500 | |
| Polyclonal | |
| Rabbit | |
| Angiogenesis, Cancer, Cellular Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, HIF Target Genes, Hypoxia, Signal Transduction, Stem Cell Markers, Tumor Suppressors, Tyrosine Kinases | |
| Tris-Glycine, 0.15 M NaCl with 0.05% Sodium Azide | |
| 3791 | |
| A synthetic peptide made to a C-terminal region of the mouse VEGF Receptor 2 protein (between residues 1300-1367). [Swiss-Prot# P35918] | |
| Primary | |
| Store at 4C. Do not freeze. | |
| 150 kDa |
| 2.55 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat | |
| CD309, CD309 antigen, EC 2.7.10, EC 2.7.10.1, Fetal liver kinase 1, fetal liver kinase-1, FLK-1, FLK1tyrosine kinase growth factor receptor, Kinase insert domain receptor, kinase insert domain receptor (a type III receptor tyrosine kinase), Protein-tyrosine kinase receptor flk-1, soluble VEGFR2, vascular endothelial growth factor receptor 2, VEGFR, VEGFR2, VEGFR-2 | |
| KDR | |
| IgG | |
| Affinity Purified | |
| Human and mouse. Predicted to react with rat based on 100% sequence homology. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title