Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    VEGFR2/KDR/Flk-1 Antibody, Alexa Fluor™ 488, Novus Biologicals™
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100529AF488
Description
VEGFR2/KDR/Flk-1 Polyclonal specifically detects VEGFR2/KDR/Flk-1 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| VEGFR2/KDR/Flk-1 | |
| Polyclonal | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| CD309, CD309 antigen, EC 2.7.10, EC 2.7.10.1, Fetal liver kinase 1, fetal liver kinase-1, FLK-1, FLK1tyrosine kinase growth factor receptor, Kinase insert domain receptor, kinase insert domain receptor (a type III receptor tyrosine kinase), Protein-tyrosine kinase receptor flk-1, soluble VEGFR2, vascular endothelial growth factor receptor 2, VEGFR, VEGFR2, VEGFR-2 | |
| Rabbit | |
| 150 kDa | |
| 0.1 mL | |
| Angiogenesis, Cancer, Cellular Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, HIF Target Genes, Hypoxia, Phospho Specific, Signal Transduction, Stem Cell Markers, Tumor Suppressors, Tyrosine Kinases | |
| 3791 | |
| Human, Mouse | |
| IgG | 
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| KDR | |
| A synthetic peptide made to an internal region of the mouse VEGF Receptor 2 protein (between residues 1200-1300). [Swiss-Prot: P35918] | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human and mouse. Immunogen sequence has 100% homology to rat. | |
| Store at 4C in the dark. | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            For Research Use Only
Spot an opportunity for improvement?Share a Content Correction