Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VEGFR3/Flt-4 Antibody (9D9) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. p-7199349 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.02 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018382 0.02 mg
NBP118651 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018382 Supplier Novus Biologicals Supplier No. NBP1186510.02MG
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

VEGFR3/Flt-4 Monoclonal specifically detects VEGFR3/Flt-4 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, CyTOF-ready.

Specifications

Antigen VEGFR3/Flt-4
Applications Immunofluorescence, Immunohistochemistry, CyTOF, Western Blot, Immunocytochemistry, Flow Cytometry, ELISA
Classification Monoclonal
Clone 9D9
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1-5ug/ml, Flow Cytometry 1-5 ug/ml, ELISA 1-5ug/ml, Immunohistochemistry 1-10ug/ml, Immunocytochemistry/Immunofluorescence 1-10ug/ml, CyTOF-ready
Formulation Sterile PBS with No Preservative
Gene Alias EC 2.7.10, EC 2.7.10.1, FLT-4, fms-related tyrosine kinase 4, LMPH1A, PCLFLT41, soluble VEGFR3 variant 1, soluble VEGFR3 variant 2, soluble VEGFR3 variant 3, Tyrosine-protein kinase receptor FLT4, vascular endothelial growth factor receptor 3, VEGFR-3, VEGFR3Fms-like tyrosine kinase 4
Gene Symbols FLT4
Host Species Mouse
Immunogen human recombinant CXCR1
Purification Method Protein A purified
Quantity 0.02 mg
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Hypoxia, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 2324
Test Specificity Mouse Monoclonal to VEGFR-3 (#1)
Reconstitution Reconstitute the antibody with 500 ul sterile PBS and the final concentration is 200 ug/ml.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.