Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VAI DECON-QUAT 200V

Catalog No. p-4591650
Click to view available options
Volume (Metric):
3.8 L
19 L
112 L
This item is not returnable. View return policy
This item is not returnable. View return policy

DECON-QUAT 200V is effective as a broad spectrum disinfectant, virucide, non-food contact sanitizer and fungicide.

  • Phosphate-free detergent
  • Concentrated formulation
  • Gamma-irradiated
  • Free rinsing
  • Neutral pH
  • Traceable
  • Applications: For use in Veterinary, Dairy, Equine, Poultry Plants, Poultry Farm, Hospitals, Restaurant, Food Handling and Process areas, Households, Hotels and Motels, Commercial, Institutional and Industrial Use

Certifications

EPA Registered Disinfectant, Virucide and Fungicide

Specifications

pH Neutral
Appearance Clear
Certifications/Compliance EPA
For Use With (Application) Effective against HIV, HBV, Salmonella, Psuedomonas, used on hard, non porous surfaces
Volume (English) 5 gal.
Volume (Metric) 19 L
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.