Description
- Regulator, complete unit and fittings for regulating Core2Clean incoming air pressure
- Pressure Relief Valve Assembly, automatic safety relief valve, factory set to prevent over pressurization of Core2Clean tank
- Stainless Steel
Specifications
Specifications
| Product Type | Clamp |
| For Use With (Equipment) | Core2Clean Plus Systems |
| For Use With (Application) | Bayonet Style Map Frame |